accessibility menu, dialog, popup

UserName

CAS RN 5989-54-8

H3CCH2(S)CH3

CAS RN 5989-54-8

IUPAC Name: (4S)-1-methyl-4-(prop-1-en-2-yl)cyclohex-1-ene
Synonyms: (-)-limonene β-limonene 4alphah-p-mentha-1,8-diene (.+-.)-limonene
Molecular Weight (g/mol): 136.24
Molecular Formula: C10H16
InChi Key: XMGQYMWWDOXHJM-SNVBAGLBSA-N
SMILES: CC(=C)[C@H]1CCC(C)=CC1
Molecular Weight (g/mol) 
  • (2)
  • (2)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
Boiling Point 
  • (2)
  • (2)
Percent Purity 
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)