CAS RN 5996-17-8
CAS RN 5996-17-8
IUPAC Name:
dipotassium (2R,3S,4R,5R)-2,3,4,5-tetrahydroxy-6-(phosphonatooxy)hexanal
Synonyms:
dipotassium D-glucose 6-phosphate
Molecular Weight (g/mol):
336.32
Molecular Formula:
C6H11K2O9P
InChi Key:
BWHWCIODKVRLNE-FAOVPRGRSA-L
SMILES:
[K+].[K+].O[C@H](COP([O-])([O-])=O)[C@@H](O)[C@H](O)[C@@H](O)C=O