CAS RN 600-90-8
CAS RN 600-90-8
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
11-epi-Prednisolone, TRC
CAS: 600-90-8 Molecular Formula: C21 H28 O5 Molecular Weight (g/mol): 360.44 Synonym: (11α)-11,17,21-Trihydroxypregna-1,4-diene-3,20-dione,Pregna-1,4-diene-3,20-dione, 11α,17,21-trihydroxy- (8CI),11α,17,21-Trihydroxy-1,4-pregnadiene-3,20-dione,Epiprednisolone,11-epi-Prednisolone IUPAC Name: (8S,9S,10R,11R,13S,14S,17R)-11,17-dihydroxy-17-(2-hydroxyacetyl)-10,13-dimethyl-7,8,9,11,12,14,15,16-octahydro-6H-cyclopenta[a]phenanthren-3-one SMILES: C[C@]12C[C@@H](O)[C@H]3[C@@H](CCC4=CC(=O)C=C[C@]34C)[C@@H]1CC[C@]2(O)C(=O)CO