accessibility menu, dialog, popup

UserName

CH3SS

CAS RN 6007-26-7

IUPAC Name: 2-methyl-1,3-dithiane
Synonyms: m-dithiane, 2-methyl-
Molecular Weight (g/mol): 134.26
Molecular Formula: C5H10S2
InChi Key: KXROTPXCYDXGSC-UHFFFAOYSA-N
SMILES: CC1SCCCS1
Molecular Weight (g/mol) 
  • (4)
  • (1)
Percent Purity 
  • (1)
  • (1)
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
Boiling Point 
  • (1)
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (4)
  • (1)
  • (1)
  • (1)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (3)
  • (2)