accessibility menu, dialog, popup

UserName

CAS RN 6009-50-3

H3CHNNOHNSCH3OO

CAS RN 6009-50-3

IUPAC Name: N-{4-[2-methyl-4-(phenylamino)-1,2,3,4-tetrahydroquinoline-1-carbonyl]phenyl}methanesulfonamide
Synonyms: N-{4-[2-methyl-4-(phenylamino)-3,4-dihydro-2H-quinoline-1-carbonyl]phenyl}methanesulfonamide
Molecular Weight (g/mol): 435.54
Molecular Formula: C24H25N3O3S
InChi Key: KCIJVZDZECUOJA-UHFFFAOYNA-N
SMILES: CC1CC(NC2=CC=CC=C2)C2=CC=CC=C2N1C(=O)C1=CC=C(NS(C)(=O)=O)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results