accessibility menu, dialog, popup

UserName

H3COOH3COH3COHNOHNO(E)

CAS RN 6015-57-2

IUPAC Name: N-{4-[(2E)-2,3-dihydro-1,3-benzoxazol-2-ylidene]-3-oxocyclohexa-1,5-dien-1-yl}-3,4,5-triethoxybenzamide
Synonyms: N-{4-[(2E)-3H-1,3-benzoxazol-2-ylidene]-3-oxocyclohexa-1,5-dien-1-yl}-3,4,5-triethoxybenzamide
Molecular Weight (g/mol): 462.5
Molecular Formula: C26H26N2O6
InChi Key: HDSCRNRNYYEKME-NLRVBDNBSA-N
SMILES: CCOC1=CC(=CC(OCC)=C1OCC)C(=O)NC1=CC(=O)\C(C=C1)=C1/NC2=CC=CC=C2O1
Quantity 
  • (2)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results