accessibility menu, dialog, popup

UserName

HNOOHOF(E/Z)

CAS RN 60252-79-1

IUPAC Name: 3-[(4-fluorophenyl)carbamoyl]prop-2-enoic acid
Synonyms: 3-[(4-fluorophenyl)carbamoyl]prop-2-enoic acid
Molecular Weight (g/mol): 209.18
Molecular Formula: C10H8FNO3
InChi Key: NRDZVHHPNZDWRA-UHFFFAOYSA-N
SMILES: OC(=O)C=CC(=O)NC1=CC=C(F)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results