accessibility menu, dialog, popup

UserName

ONOOHNO

CAS RN 603-62-3

IUPAC Name: 4-nitro-2,3-dihydro-1H-isoindole-1,3-dione
Synonyms: 3-nitrophthalimide
Molecular Weight (g/mol): 192.13
Molecular Formula: C8H4N2O4
InChi Key: BONIIQYTWOPUQI-UHFFFAOYSA-N
SMILES: [O-][N+](=O)C1=CC=CC2=C1C(=O)NC2=O
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results