CAS RN 603-99-6
CAS RN 603-99-6
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Pregnanetriolone, TRC
CAS: 603-99-6 Molecular Formula: C21H34O4 Molecular Weight (g/mol): 350.49 Synonym: 3α,17,20α-Trihydroxy-5β-pregnan-11-one,11-keto-Pregnane-3α,17,20α-triol,3α,17α,20α-Trihydroxy-5β-pregnan-11-one,5β-Pregnan-3α,17α,20α-triol-11-one,5β-Pregnane-3α,17α,20α-triol-11-one,NSC 79097 IUPAC Name: (3R,5R,8S,9S,10S,13S,14S,17R)-3,17-dihydroxy-17-[(1S)-1-hydroxyethyl]-10,13-dimethyl-2,3,4,5,6,7,8,9,12,14,15,16-dodecahydro-1H-cyclopenta[a]phenanthren-11-one SMILES: C[C@H](O)[C@@]1(O)CC[C@H]2[C@@H]3CC[C@@H]4C[C@H](O)CC[C@]4(C)[C@H]3C(=O)C[C@]12C