CAS RN 60398-91-6
CAS RN 60398-91-6
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Bufuralol Hydrochloride, TRC
CAS: 60398-91-6 Molecular Formula: C16 H23 N O2 . Cl H Molecular Weight (g/mol): 297.82 IUPAC Name: 2-(tert-butylamino)-1-(7-ethyl-1-benzofuran-2-yl)ethanol;hydrochloride SMILES: Cl.CCc1cccc2cc(oc12)C(O)CNC(C)(C)C