accessibility menu, dialog, popup

UserName

CAS RN 60404-18-4

ClSClBr

CAS RN 60404-18-4

IUPAC Name: 3-bromo-2,5-dichlorothiophene
Synonyms: 3-bromo-2,5-dichlorothiophene
Molecular Weight (g/mol): 231.92
Molecular Formula: C4HBrCl2S
InChi Key: PBUHOXBSIQJRNO-UHFFFAOYSA-N
SMILES: ClC1=CC(Br)=C(Cl)S1
Boiling Point 
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (3)
  • (3)
  • (1)
  • (1)
  • (1)