accessibility menu, dialog, popup

UserName

H3COO(R)O

CAS RN 60456-26-0

IUPAC Name: [(2R)-oxiran-2-yl]methyl butanoate
Synonyms: (2R)-oxiran-2-ylmethyl butanoate
Molecular Weight (g/mol): 144.17
Molecular Formula: C7H12O3
InChi Key: YLNSNVGRSIOCEU-ZCFIWIBFSA-N
SMILES: CCCC(=O)OC[C@H]1CO1
Quantity 
  • (1)
  • (1)
  • (2)
  • (1)
Boiling Point 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results