accessibility menu, dialog, popup

UserName

CAS RN 60459-08-7

OSOOOCo

CAS RN 60459-08-7

IUPAC Name: λ²-cobalt(2+) hydrate sulfate
Synonyms: λ²-cobalt(2+) hydrate sulfate
Molecular Weight (g/mol): 173
Molecular Formula: CoH2O5S
InChi Key: BGORGFZEVHFAQU-UHFFFAOYSA-L
SMILES: O.[Co++].[O-]S([O-])(=O)=O
Grade 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results