accessibility menu, dialog, popup

UserName

CAS RN 6051-62-3

H2NNHOHH2NNO(Z)(Z)

CAS RN 6051-62-3

IUPAC Name: 1-[(1Z,4Z)-4-[amino(hydroxyamino)methylidene]cyclohexa-2,5-dien-1-ylidene]-1-nitrosomethanamine
Synonyms: 1-[(1Z,4Z)-4-[amino(hydroxyamino)methylidene]cyclohexa-2,5-dien-1-ylidene]-1-nitrosomethanamine
Molecular Weight (g/mol): 194.19
Molecular Formula: C8H10N4O2
InChi Key: JUXCYRYHZGAKKG-SFECMWDFSA-N
SMILES: NC(NO)=C1C=CC(C=C1)=C(N)N=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results