accessibility menu, dialog, popup

UserName

HNNOOOSOOONaNa(E/Z)

CAS RN 6054-99-5

IUPAC Name: disodium 6-oxo-3-[2-(4-sulfonatophenyl)hydrazin-1-ylidene]cyclohexa-1,4-diene-1-carboxylate
Synonyms: disodium 6-oxo-3-[2-(4-sulfonatophenyl)hydrazin-1-ylidene]cyclohexa-1,4-diene-1-carboxylate
Molecular Weight (g/mol): 366.25
Molecular Formula: C13H8N2Na2O6S
InChi Key: CWQLKNYZZSWCNX-UHFFFAOYSA-L
SMILES: [Na+].[Na+].[O-]C(=O)C1=CC(C=CC1=O)=NNC1=CC=C(C=C1)S([O-])(=O)=O
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results