CAS RN 60599-38-4
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
N-Nitrosobis(2-oxopropyl)amine, TRC
CAS: 60599-38-4 Molecular Formula: C6 H10 N2 O3 Molecular Weight (g/mol): 158.16 Synonym: N-Nitroso Bis(2-Oxopropyl)amine IUPAC Name: N,N-bis(2-oxopropyl)nitrous amide SMILES: CC(=O)CN(CC(=O)C)N=O