CAS RN 6068-90-2
IUPAC Name:
(isothiocyanatomethyl)cyclopropane
Synonyms:
(isothiocyanatomethyl)cyclopropane
Molecular Weight (g/mol):
113.18
Molecular Formula:
C5H7NS
InChi Key:
YRJONJBJAYHHQB-UHFFFAOYSA-N
SMILES:
S=C=NCC1CC1
1
–
1
of
1
results
1
Cyclopropylmethyleisothiocyanate, 97%, Thermo Scientific™
CAS: 6068-90-2 Molecular Formula: C5H7NS Molecular Weight (g/mol): 113.178 MDL Number: MFCD00052446 InChI Key: YRJONJBJAYHHQB-UHFFFAOYSA-N Synonym: cyclopropylmethyl isothiocyanate,isothiocyanatomethyl cyclopropane,cyclopropylmethylisothiocyanate,cyclopropane, isothiocyanatomethyl,cyclopropylmethanisothiocyanate PubChem CID: 2795403 IUPAC Name: isothiocyanatomethylcyclopropane SMILES: C1CC1CN=C=S