accessibility menu, dialog, popup

UserName

CAS RN 608-68-4

H3COO(S)OH(R)OHOOCH3

CAS RN 608-68-4

IUPAC Name: 1,4-dimethyl (2R,3S)-2,3-dihydroxybutanedioate
Synonyms: 1,4-dimethyl (2R,3S)-2,3-dihydroxybutanedioate
Molecular Weight (g/mol): 178.14
Molecular Formula: C6H10O6
InChi Key: PVRATXCXJDHJJN-ZXZARUISSA-N
SMILES: COC(=O)[C@@H](O)[C@@H](O)C(=O)OC
Boiling Point 
  • (3)
  • (1)
Percent Purity 
  • (3)
  • (1)
Quantity 
  • (2)
  • (1)
  • (1)
Physical Form 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (1)
  • (3)
  • (1)
  • (3)
  • (1)
  • (2)
  • (1)
  • (1)
  • (3)