accessibility menu, dialog, popup

UserName

H3CFOHO

CAS RN 6087-13-4

IUPAC Name: 2-fluoropropanoic acid
Synonyms: 2-fluoropropanoic acid
Molecular Weight (g/mol): 92.07
Molecular Formula: C3H5FO2
InChi Key: ZVZPFTCEXIGSHM-UHFFFAOYNA-N
SMILES: CC(F)C(O)=O
Molecular Weight (g/mol) 
  • (2)
  • (2)
Quantity 
  • (2)
  • (1)
  • (1)
Boiling Point 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)
  • (2)