accessibility menu, dialog, popup

UserName

H2NOO

CAS RN 609-38-1

IUPAC Name: furan-2-carboxamide
Synonyms: furamide
Molecular Weight (g/mol): 111.1
Molecular Formula: C5H5NO2
InChi Key: TVFIYRKPCACCNL-UHFFFAOYSA-N
SMILES: NC(=O)C1=CC=CO1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results