accessibility menu, dialog, popup

UserName

CAS RN 61226-19-5

IUPAC Name: 3,5-dimethyl-1-phenyl-1H-pyrazole-4-carboxylate
Synonyms: 3,5-dimethyl-1-phenylpyrazole-4-carboxylate
Molecular Weight (g/mol): 215.23
Molecular Formula: C12H11N2O2
InChi Key: LPYTYYLNGJGJGW-UHFFFAOYSA-M
SMILES: CC1=NN(C(C)=C1C([O-])=O)C1=CC=CC=C1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results