accessibility menu, dialog, popup

UserName

CAS RN 612845-44-0

BNOCH3HOOH

CAS RN 612845-44-0

IUPAC Name: (6-ethoxypyridin-3-yl)boronic acid
Synonyms: 6-ethoxypyridin-3-ylboronic acid
Molecular Weight (g/mol): 166.97
Molecular Formula: C7H10BNO3
InChi Key: UONCERAQKBPLML-UHFFFAOYSA-N
SMILES: CCOC1=NC=C(C=C1)B(O)O
Molecular Weight (g/mol) 
  • (2)
  • (3)
Percent Purity 
  • (2)
  • (3)
Quantity 
  • (2)
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (3)
  • (2)
  • (3)
  • (2)
  • (3)
  • (2)
  • (1)
  • (2)