accessibility menu, dialog, popup

UserName

CAS RN 61291-21-2

IUPAC Name: 5-methyl-1,3-thiazole-2-carboxylic acid
Synonyms: 5-methyl-1,3-thiazole-2-carboxylic acid
Molecular Weight (g/mol): 143.16
Molecular Formula: C5H5NO2S
InChi Key: UYDGNRLMYZMDOG-UHFFFAOYSA-N
SMILES: CC1=CN=C(S1)C(O)=O
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results