accessibility menu, dialog, popup

UserName

CAS RN 613-35-4

CH3ONHNHCH3O

CAS RN 613-35-4

IUPAC Name: N-{4'-acetamido-[1,1'-biphenyl]-4-yl}acetamide
Synonyms: N,N'-diacetylbenzidine
Molecular Weight (g/mol): 268.32
Molecular Formula: C16H16N2O2
InChi Key: CZVHCFKUXGRABC-UHFFFAOYSA-N
SMILES: CC(=O)NC1=CC=C(C=C1)C1=CC=C(NC(C)=O)C=C1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results