CAS RN 6132-02-1
IUPAC Name:
disodium decahydrate carbonate
Synonyms:
disodium decahydrate carbonate
Molecular Weight (g/mol):
286.14
Molecular Formula:
CH20Na2O13
InChi Key:
XYQRXRFVKUPBQN-UHFFFAOYSA-L
SMILES:
O.O.O.O.O.O.O.O.O.O.[Na+].[Na+].[O-]C([O-])=O
1
–
1
of
1
results
1
Promotions Available
Sodium carbonate decahydrate, 99+%, for analysis
Sodium carbonate decahydrate, 99+%, for analysis
CAS: 6132-02-1 Molecular Formula: CNa2O3·10H2O Molecular Weight (g/mol): 286.14 MDL Number: MFCD00149178 InChI Key: XYQRXRFVKUPBQN-UHFFFAOYSA-L Synonym: sodium carbonate decahydrate,natrii carbonas decahydricus,carbonic acid disodium salt, decahydrate,unii-ls505bg22i,disodium decahydrate carbonate,acmc-1b9jt,natrium-carbonat-dekahydrat,natrium-carbonat-10-wasser,disodium carbonate decahydrate,ksc353a3t PubChem CID: 151402 IUPAC Name: disodium;carbonate;decahydrate SMILES: C(=O)([O-])[O-].O.O.O.O.O.O.O.O.O.O.[Na+].[Na+]