accessibility menu, dialog, popup

UserName

CAS RN 613239-76-2

IUPAC Name: 2-[4-(chloromethyl)phenyl]-5-(trifluoromethyl)pyridine
Synonyms: 2-[4-(chloromethyl)phenyl]-5-(trifluoromethyl)pyridine
Molecular Weight (g/mol): 271.67
Molecular Formula: C13H9ClF3N
InChi Key: JTXHHQDYEHXWPC-UHFFFAOYSA-N
SMILES: FC(F)(F)C1=CN=C(C=C1)C1=CC=C(CCl)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results