accessibility menu, dialog, popup

UserName

H2NOHNO

CAS RN 614-22-2

IUPAC Name: benzoylurea
Synonyms: benzoylurea ureide benzoic acid benzoic acid, ureide
Molecular Weight (g/mol): 164.16
Molecular Formula: C8H8N2O2
InChi Key: HRYILSDLIGTCOP-UHFFFAOYSA-N
SMILES: NC(=O)NC(=O)C1=CC=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results