accessibility menu, dialog, popup

UserName

H2NSHNO

CAS RN 614-23-3

IUPAC Name: benzoylthiourea
Synonyms: benzoylthiourea
Molecular Weight (g/mol): 180.23
Molecular Formula: C8H8N2OS
InChi Key: DQMWMUMCNOJLSI-UHFFFAOYSA-N
SMILES: NC(=S)NC(=O)C1=CC=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results