accessibility menu, dialog, popup

UserName

CH3OOOH3C

CAS RN 6141-58-8

IUPAC Name: methyl 2-methylfuran-3-carboxylate
Synonyms: methyl 2-methylfuran-3-carboxylate
Molecular Weight (g/mol): 140.14
Molecular Formula: C7H8O3
InChi Key: UVRRIABXNIGUJZ-UHFFFAOYSA-N
SMILES: COC(=O)C1=C(C)OC=C1
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (1)
  • (1)