accessibility menu, dialog, popup

UserName

NNFFF

CAS RN 61655-67-2

IUPAC Name: 2-(trifluoromethyl)pyrazine
Synonyms: 2-(trifluoromethyl)pyrazine
Molecular Weight (g/mol): 148.09
Molecular Formula: C5H3F3N2
InChi Key: CEAMSISEJZMQEP-UHFFFAOYSA-N
SMILES: FC(F)(F)C1=NC=CN=C1
Boiling Point 
  • (2)
  • (2)
Molecular Weight (g/mol) 
  • (2)
  • (2)
Quantity 
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)