accessibility menu, dialog, popup

UserName

NaOO

CAS RN 61886-29-1

IUPAC Name: sodium 4-cyclohexylbutanoate
Synonyms: sodium 4-cyclohexylbutanoate
Molecular Weight (g/mol): 192.23
Molecular Formula: C10H17NaO2
InChi Key: GBEFSJZAXXWKIO-UHFFFAOYSA-M
SMILES: [Na+].[O-]C(=O)CCCC1CCCCC1
Grade 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results