accessibility menu, dialog, popup

UserName

CAS RN 61888-78-6

IUPAC Name: 4-(2,4-dichlorophenyl)but-3-en-2-one
Synonyms: 4-(2,4-dichlorophenyl)but-3-en-2-one
Molecular Weight (g/mol): 215.07
Molecular Formula: C10H8Cl2O
InChi Key: SAEQHTBHPNKKRX-UHFFFAOYSA-N
SMILES: CC(=O)C=CC1=C(Cl)C=C(Cl)C=C1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results