accessibility menu, dialog, popup

UserName

CAS RN 620-08-6

H3CON

CAS RN 620-08-6

IUPAC Name: 4-methoxypyridine
Synonyms: 4-methoxypyridine
Molecular Weight (g/mol): 109.13
Molecular Formula: C6H7NO
InChi Key: XQABVLBGNWBWIV-UHFFFAOYSA-N
SMILES: COC1=CC=NC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results