accessibility menu, dialog, popup

UserName

H2NSHN

CAS RN 621-83-0

IUPAC Name: benzylthiourea
Synonyms: benzylthiourea
Molecular Weight (g/mol): 166.24
Molecular Formula: C8H10N2S
InChi Key: UCGFRIAOVLXVKL-UHFFFAOYSA-N
SMILES: NC(=S)NCC1=CC=CC=C1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results