accessibility menu, dialog, popup

UserName

NNN

CAS RN 622-79-7

IUPAC Name: (azidomethyl)benzene
Synonyms: (azidomethyl)benzene
Molecular Weight (g/mol): 133.15
Molecular Formula: C7H7N3
InChi Key: UDLLFLQFQMACJB-UHFFFAOYSA-N
SMILES: [N-]=[N+]=NCC1=CC=CC=C1
Boiling Point 
  • (1)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (3)
  • (1)
  • (1)
  • (1)