accessibility menu, dialog, popup

UserName

CAS RN 623-15-4

H3COO(E)

CAS RN 623-15-4

IUPAC Name: (3E)-4-(furan-2-yl)but-3-en-2-one
Synonyms: 3-buten-2-one, 4-(2-furanyl)-
Molecular Weight (g/mol): 136.15
Molecular Formula: C8H8O2
InChi Key: GBKGJMYPQZODMI-SNAWJCMRSA-N
SMILES: CC(=O)\C=C\C1=CC=CO1
Quantity 
  • (1)
  • (2)
  • (2)
  • (1)
  • (2)
  • (1)
Percent Purity 
  • (3)
  • (3)
Boiling Point 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (3)
  • (1)
  • (2)
  • (2)
  • (1)
  • (2)
  • (1)
  • (3)
  • (3)
  • (3)