accessibility menu, dialog, popup

UserName

H3COON

CAS RN 623-49-4

IUPAC Name: ethyl cyanoformate
Synonyms: carbonocyanidic acid, ethyl ester
Molecular Weight (g/mol): 99.09
Molecular Formula: C4H5NO2
InChi Key: MSMGXWFHBSCQFB-UHFFFAOYSA-N
SMILES: CCOC(=O)C#N
Quantity 
  • (2)
  • (1)
  • (2)
  • (1)
  • (2)
Boiling Point 
  • (5)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (3)
  • (3)
  • (2)
  • (1)
  • (2)
  • (1)
  • (2)
  • (5)
  • (2)