accessibility menu, dialog, popup

UserName

CAS RN 623-61-0

IUPAC Name: propan-2-yl 2-hydroxyacetate
Synonyms: isopropyl 2-hydroxyacetate
Molecular Weight (g/mol): 118.13
Molecular Formula: C5H10O3
InChi Key: AZKIQQBSVTWCGY-UHFFFAOYSA-N
SMILES: CC(C)OC(=O)CO

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results