accessibility menu, dialog, popup

UserName

CAS RN 6230-11-1

(S)ONH2OHOH3C

CAS RN 6230-11-1

IUPAC Name: 2-amino-3-(4-methoxyphenyl)propanoic acid
Synonyms: 2-amino-3-(4-methoxyphenyl)propanoic acid
Molecular Weight (g/mol): 195.22
Molecular Formula: C10H13NO3
InChi Key: GEYBMYRBIABFTA-UHFFFAOYNA-N
SMILES: COC1=CC=C(CC(N)C(O)=O)C=C1
Molecular Weight (g/mol) 
  • (1)
  • (7)
Quantity 
  • (3)
  • (1)
  • (1)
  • (3)
Percent Purity 
  • (3)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (2)
  • (3)
  • (1)
  • (7)
  • (3)
  • (1)
  • (1)
  • (3)
  • (3)
  • (2)