accessibility menu, dialog, popup

UserName

H3COOCH3O

CAS RN 624-45-3

IUPAC Name: methyl 4-oxopentanoate
Synonyms: methyl 4-oxopentanoate
Molecular Weight (g/mol): 130.14
Molecular Formula: C6H10O3
InChi Key: UAGJVSRUFNSIHR-UHFFFAOYSA-N
SMILES: COC(=O)CCC(C)=O
Molecular Weight (g/mol) 
  • (7)
  • (3)
Quantity 
  • (1)
  • (3)
  • (3)
  • (1)
  • (2)
Percent Purity 
  • (3)
  • (4)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (4)
  • (3)
  • (3)
  • (4)
  • (7)
  • (3)
  • (1)
  • (3)
  • (3)
  • (1)
  • (2)
  • (3)
  • (4)
  • (3)
  • (3)