accessibility menu, dialog, popup

UserName

CAS RN 624-70-4

IUPAC Name: 1-chloro-2-iodoethane
Synonyms: 1-chloro-2-iodoethane
Molecular Weight (g/mol): 190.41
Molecular Formula: C2H4ClI
InChi Key: JTWWWQGSFTWWDL-UHFFFAOYSA-N
SMILES: ClCCI
Boiling Point 
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results