accessibility menu, dialog, popup

UserName

CAS RN 625-51-4

CH3OHNHO

CAS RN 625-51-4

IUPAC Name: N-(hydroxymethyl)acetamide
Synonyms: formicin
Molecular Weight (g/mol): 89.09
Molecular Formula: C3H7NO2
InChi Key: HWJHZLJIIWOTGZ-UHFFFAOYSA-N
SMILES: CC(=O)NCO
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results