accessibility menu, dialog, popup

UserName

CAS RN 625-69-4

CH3HOCH3HO

CAS RN 625-69-4

IUPAC Name: pentane-2,4-diol
Synonyms: 2,4-pentanediol
Molecular Weight (g/mol): 104.15
Molecular Formula: C5H12O2
InChi Key: GTCCGKPBSJZVRZ-UHFFFAOYNA-N
SMILES: CC(O)CC(C)O
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (1)
  • (1)