accessibility menu, dialog, popup

UserName

OWOOOOOCH3H3CH3CCH3CH3CH3

CAS RN 62571-53-3

IUPAC Name: hexaethanolate tungsten
Synonyms: hexakis(ethoxide) tungsten
Molecular Weight (g/mol): 454.21
Molecular Formula: C12H30O6W
InChi Key: YJBCFLKOYPGFTR-UHFFFAOYSA-N
SMILES: [W].CC[O-].CC[O-].CC[O-].CC[O-].CC[O-].CC[O-]
Physical Form 
  • (2)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)