accessibility menu, dialog, popup

UserName

OHOHN

CAS RN 626-03-9

IUPAC Name: 2-hydroxy-1,4-dihydropyridin-4-one
Synonyms: 2-hydroxy-1H-pyridin-4-one
Molecular Weight (g/mol): 111.1
Molecular Formula: C5H5NO2
InChi Key: ZEZJPIDPVXJEME-UHFFFAOYSA-N
SMILES: OC1=CC(=O)C=CN1
Quantity 
  • (2)
  • (1)
  • (2)
  • (3)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results