accessibility menu, dialog, popup

UserName

CAS RN 626-35-7

H3COONOO

CAS RN 626-35-7

IUPAC Name: ethyl 2-nitroacetate
Synonyms: nitroacetic acid ethyl ester
Molecular Weight (g/mol): 133.1
Molecular Formula: C4H7NO4
InChi Key: FTKASJMIPSSXBP-UHFFFAOYSA-N
SMILES: CCOC(=O)C[N+]([O-])=O
Molecular Weight (g/mol) 
  • (6)
  • (4)
Quantity 
  • (1)
  • (1)
  • (2)
  • (3)
  • (3)
Boiling Point 
  • (3)
  • (4)
Percent Purity 
  • (4)
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (4)
  • (3)
  • (6)
  • (4)
  • (1)
  • (1)
  • (2)
  • (3)
  • (3)
  • (3)
  • (4)
  • (4)
  • (3)
  • (3)