CAS RN 62625-16-5
CAS RN 62625-16-5
IUPAC Name:
tripotassium 2-{hydroxybis[4-(sulfonatooxy)phenyl]methyl}benzoate
Synonyms:
tripotassium 2-{hydroxybis[4-(sulfonatooxy)phenyl]methyl}benzoate
Molecular Weight (g/mol):
610.73
Molecular Formula:
C20H13K3O11S2
InChi Key:
PLHSVCWOEHYSHO-UHFFFAOYSA-K
SMILES:
[K+].[K+].[K+].OC(C1=CC=C(OS([O-])(=O)=O)C=C1)(C1=CC=C(OS([O-])(=O)=O)C=C1)C1=CC=CC=C1C([O-])=O
1
–
2
of
2
results
1
Thermo Scientific Chemicals Phenolphthalein disulfate tripotassium salt hydrate, 98%
CAS: 62625-16-5 Molecular Formula: C20H13K3O11S2 Molecular Weight (g/mol): 610.73 MDL Number: MFCD00150194 InChI Key: PLHSVCWOEHYSHO-UHFFFAOYSA-K Synonym: phenolphthalein disulfate potassium salt,phenolphthalein disulfate tripotassium salt hydrate PubChem CID: 129893851 SMILES: [K+].[K+].[K+].OC(C1=CC=C(OS([O-])(=O)=O)C=C1)(C1=CC=C(OS([O-])(=O)=O)C=C1)C1=CC=CC=C1C([O-])=O
2
Thermo Scientific Chemicals Phenolphthalein disulfate, tripotassium salt trihydrate, 98%, pure
CAS: 62625-16-5 Molecular Formula: C20H13K3O11S2 Molecular Weight (g/mol): 610.73 MDL Number: MFCD00150194 InChI Key: PLHSVCWOEHYSHO-UHFFFAOYSA-K PubChem CID: 129893851 SMILES: [K+].[K+].[K+].OC(C1=CC=C(OS([O-])(=O)=O)C=C1)(C1=CC=C(OS([O-])(=O)=O)C=C1)C1=CC=CC=C1C([O-])=O