accessibility menu, dialog, popup

UserName

PbOOOO

CAS RN 62637-99-4

IUPAC Name: λ²-lead(2+) bis(4-cyclohexylbutanoate)
Synonyms: λ²-lead(2+) bis(4-cyclohexylbutanoate)
Molecular Weight (g/mol): 545.7
Molecular Formula: C20H34O4Pb
InChi Key: CCJZHCZMNGUOCA-UHFFFAOYSA-L
SMILES: [Pb++].[O-]C(=O)CCCC1CCCCC1.[O-]C(=O)CCCC1CCCCC1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results