CAS RN 627-20-3
IUPAC Name:
(2Z)-pent-2-ene
Synonyms:
cis-2-pentene
Molecular Weight (g/mol):
70.14
Molecular Formula:
C5H10
InChi Key:
QMMOXUPEWRXHJS-HYXAFXHYSA-N
SMILES:
CC\C=C/C
1
–
1
of
1
results
1
cis-2-Pentene
CAS: 627-20-3 Molecular Formula: C5H10 Molecular Weight (g/mol): 70.14 MDL Number: MFCD00063978 InChI Key: QMMOXUPEWRXHJS-HYXAFXHYSA-N Synonym: cis-2-pentene,z-2-pentene,2-pentene, z,cis-pentene,z-pent-2-ene,unii-54ur3xz4fc,cis-pentene-2,54ur3xz4fc,2-pentene,2-pentene, 2z PubChem CID: 5326160 IUPAC Name: (Z)-pent-2-ene SMILES: CC\C=C/C