accessibility menu, dialog, popup

UserName

CH3NCS

CAS RN 628-30-8

IUPAC Name: 1-isothiocyanatopropane
Synonyms: propane, 1-isothiocyanato-
Molecular Weight (g/mol): 101.17
Molecular Formula: C4H7NS
InChi Key: KKASGUHLXWAKEZ-UHFFFAOYSA-N
SMILES: CCCN=C=S
Percent Purity 
  • (1)
  • (5)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
Boiling Point 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (1)
  • (5)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (3)